Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
anti-Nbs1, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 2 publications
Supplier: Novus Biologicals NBP106705
Description
Nbs1 Polyclonal specifically detects Nbs1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Nbs1 | |
Polyclonal | |
Unconjugated | |
PBS and 30% Glycerol with 0.1% Sodium Azide | |
NBN | |
Nbs1 Antibody was made to a synthetic peptide made to an internal portion of the mouse NBS1 protein (within residues 150-200). [Swiss-Prot# Q9R207] | |
Affinity Purified | |
RUO | |
Primary | |
Human, Mouse | |
IgG |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
1.48 mg/mL | |
Western Blot 0.5 ug/ml, Immunohistochemistry 1:100, Immunohistochemistry-Paraffin 1:100 | |
ATV, AT-V1, AT-V2, Cell cycle regulatory protein p95, FLJ10155, MGC87362, NBS, NBS1P95, nibrin, Nijmegen breakage syndrome 1 (nibrin), Nijmegen breakage syndrome protein 1, p95, p95 protein of the MRE11/RAD50 complex | |
Rabbit | |
84 kDa | |
0.1 mL | |
Breast Cancer, Cancer, Cell Cycle and Replication, Checkpoint signaling, Chromatin Research, DNA Double Strand Break Repair, DNA Repair, Homologous Recombination, Non homologous end joining, Tumor Suppressors | |
4683 | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only