Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NCAM Mouse anti-Human, Pig, Clone: ERIC-1, MilliporeSigma™

Catalog No. MAB2120ZMI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB2120ZMI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB2120ZMI Supplier MilliporeSigma Supplier No. MAB2120Z
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-NCAM Antibody, extracellular, clone ERIC-1, Azide free is an antibody against NCAM for use in ELISA, western blot, immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NCAM
Applications ELISA, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone ERIC-1
Conjugate Unconjugated
Formulation Liquid in PBS. Does not contain any preservatives
Gene Accession No. NM_000615.5; NM_001076682.2; NM_181351.3
Gene Symbols NCAM1, CD56, NCAM-140, NCAM, NCAM-120, MSK39
Host Species Mouse
Immunogen Human Retinoblastoma tumor
Quantity 100 μg
Primary or Secondary Primary
Target Species Human, Pig
Content And Storage −20°C in undiluted aliquots. Avoid repeated freeze/thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.