Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NCK Mouse, Unlabeled, Clone: 108, BD

Catalog No. BDB610099 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB610099 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB610099 Supplier BD Biosciences Supplier No. 610099
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

NCK is a 47kDa protein that belongs to a class of signaling molecules called adaptors. These proteins exert their effects by coupling activated receptors to downstream effectors. NCK is comprised almost entirely of one SH2 and three SH3 domains and it possesses no intrinsic catalytic activity. Investigations of its interactions with cellular proteins have found constitutive association of NCK with various Ser/THr kinases, such as p21-associated kinase (PAK) and casein kinase 1. Other proteins that directly interact with NCK are: Sos, Cbl, Wiskott-Aldrich syndrome protein (WASP), IRS1, and multiple tyrosine kinase receptors. The presence of NCK is necessary for the mitogenic response elicited by PDGF in some cells. In addition, the Drosophila homologue of NCK is involved in regulating cytoskeletal organization.

Host Species: Mouse
Clone: 108
Isotype: IgG2b
Species Reactivity: Human
Immunogen: Human NCK aa. 279-377
Formula Weight [Chemical]: 47kDa

Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen NCK
Applications Western Blot
Classification Monoclonal
Clone 108
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human NCK aa. 279-377
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Bovine, Canine, Chicken, Frog, Human, Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG2b
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.