Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nck Rabbit anti-Human, Mouse, Rat, Polyclonal, MilliporeSigma™

Catalog No. 06288MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
06-288-MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 06-288-MI Supplier MilliporeSigma Supplier No. 06288
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Detect Nck using this Anti-Nck Antibody validated for use in immunoprecipitation and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Nck
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation 0.1M Tris-glycine, pH 7.4, 0.15M NaCl containing 0.05% sodium azide
Gene Symbols NCK1; MGC12668; NCK; NCKalpha
Host Species Rabbit
Immunogen Purified GST fusion protein produced in E. coli., corresponding to human Nck amino acid sequence 1 to 254
Purification Method Protein G chromatography
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) NM_010878
Target Species Human, Mouse, Rat
Content And Storage −20°C for 24 months
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.