Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NCK2, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89003102 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-003-102 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-003-102 Supplier Abnova Corporation Supplier No. H00008440A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant NCK2.

This gene encodes a member of the NCK family of adaptor proteins. The protein contains three SH3 domains and one SH2 domain. The protein has no known catalytic function but has been shown to bind and recruit various proteins involved in the regulation of receptor protein tyrosine kinases. It is through these regulatory activities that this protein is believed to be involved in cytoskeletal reorganization. Alternate transcriptional splice variants, encoding different isoforms, have been characterized. [provided by RefSeq

Sequence: LYPFSSVTEEELNFEKGETMEVIEKPENDPEWWKCKNARGQVGLVPKNYVVVLSDGPALHPAHAPQISYTGPSSSGRFAGREWYYGNVTRHQAECALNERGVEGDFLIRD

Specifications

Antigen NCK2
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant NCK2.
Formulation 50% glycerol
Gene NCK2
Gene Accession No. NM_003581
Gene Alias GRB4/NCKbeta
Gene Symbols NCK2
Host Species Mouse
Immunogen NCK2 (NP_003572, 203 a.a. ∼ 312 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8440
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.