Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NCoA-2 Rabbit anti-Human, Polyclonal, MilliporeSigma™

Catalog No. 071799MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
07-179-9MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07-179-9MI Supplier MilliporeSigma Supplier No. 071799
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Use Anti-NCoA-2 Antibody (rabbit polyclonal antibody) validated in western blotting to detect NCoA-2 also known as Transcriptional intermediary factor 2, glucocorticoid receptor-interacting protein-1, nuclear receptor coactivator 2.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NCoA-2
Applications ChIP Assay, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4, 150mM NaCl) with 0.05% sodium azide.
Gene Symbols NCOA2; GRIP1; KAT13C; NCoA-2; SRC2; TIF2; bHLHe75; hTIF2; SRC-2; TIF-2
Host Species Rabbit
Immunogen Linear peptide corresponding to human NCoA-2
Purification Method Affinity Purfied
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) NP_006531.1
Target Species Human
Content And Storage 2-8°C for 12 months
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.