Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

nuclear receptor coactivator 4, Mouse, Clone: 1B7, Abnova™

Catalog No. 89001074 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-001-074 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-001-074 Supplier Abnova Corporation Supplier No. H00008031M04
Add to Cart
Edge
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant NCOA4.

This gene encodes an androgen receptor coactivator. The encoded protein interacts with the androgen receptor in a ligand-dependent manner to enhance its transcriptional activity. Chromosomal translocations between this gene and the ret tyrosine kinase gene, also located on chromosome 10, have been associated with papillary thyroid carcinoma. Alternatively spliced transcript variants have been described. Pseudogenes are present on chromosomes 4, 5, 10, and 14. [provided by RefSeq

Sequence: EGSPKEVPGTEDRAGKQKFKSPMNTSWCSFNTADWVLPGKKMGNLSQLSSGEDKWLLRKKAQEVLLNSPLQEEHNFPPDHYGLPAVCDLFACMQLKVDKEKWLYRTPLQM

Specifications

Antigen nuclear receptor coactivator 4
Applications ELISA, Immunofluorescence, Western Blot
Classification Monoclonal
Clone 1B7
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant NCOA4.
Formulation PBS with no preservative; pH 7.4
Gene NCOA4
Gene Accession No. NM_005437
Gene Alias ARA70/DKFZp762E1112/ELE1/PTC3/RFG
Gene Symbols NCOA4
Host Species Mouse
Immunogen NCOA4 (NP_005428, 505 a.a. ∼ 614 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Transcription Regulation
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 8031
Target Species Human, Mouse
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.