Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-NDE1 Clone: 2G11-1C11,

Catalog No. ST1699100UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
ST1699100UG 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ST1699100UG Supplier MilliporeSigma™ Supplier No. ST1699100UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifically detects NDE1 Clone: 2G11-1C11 in Human samples, and it is validated for ELISA, Immunocytochemistry, Immuno Blotting

This gene encodes a member of the nuclear distribution E (NudE) family of proteins. The encoded protein is localized at the centrosome and interacts with other centrosome components as part of a multiprotein complex that regulates dynein function. This protein plays an essential role in microtubule organization, mitosis and neuronal migration. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NDE1
Applications ELISA, Immunoblot, Immunoblot, Immunocytochemistry
Classification Monoclonal
Clone 2G11-1C11
Conjugate Unconjugated
Formulation In PBS, pH 7.2.
Host Species Mouse
Immunogen Full-length, recombinant, human NDE1 (aa 1-336), expressed as a GST fusion protein
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes the ∽40-45kDa NDE1 protein in HeLa cells.
Target Species Human
Content And Storage −20°C
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.