Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NADPH dependent diflavin oxidoreductase 1, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89018138 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-018-138 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-018-138 Supplier Abnova Corporation Supplier No. H00027158D01P
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against a full-length human NDOR1 protein.

This gene encodes an NADPH-dependent diflavin reductase that contains both flavin mononucleotide (FMN) and flavin adenine dinucleotide (FAD) binding domains. The encoded protein is an enzyme that catalyzes the transfers electrons from NADPH through FAD and FMN cofactors to potential redox partners. Alternative splicing results in multiple transcript variants. [provided by RefSeq

Sequence: MPSPQLLVLFGSQTGTAQDVSERLGREARRRRLGCRVQALDSYPVVNLINEPLVIFVCATTGQGDPPDNMKNFWRFIFRKNLPSTALCQMDFAVLGLGDSSYAKFNFVAKKLHRRLLQLGGSALLPVCLGDDQHELGPDAAVDPWLRDLWDRVLGLYPPPPGLTEIPPGVPLPSKFTLLFLQEAPSTGSEGQRVAHPGSQEPPSESKPFLAPMISNQRVTGPSHFQDVRLIEFDILGSGISFAAGDVVLIQPSNSAAHVQRFCQVLGLDPDQLFMLQPREPDVSSPTRLPQPCSMRHLVSHYLDIASVPRRSFFELLACLSLHELEREKLLEFSSAQGQEELFEYCNRPRRTILEVLCDFPHTAAAIPPDYLLDLIPVIRPRAFSIASSLLTHPSRLQILVAVVQFQTRLKEPRRGLCSSWLASLDPGQGPVRVPLWVRPGSLAFPETPDTPVIMVGPGTGVAPFRAAIQERVAQGQTGNFLFFGCRWRDQDFYWEAEWQELEKRDCLTLIPAFSREQEQKIYVQHRLRELGSLVWELLDRQGAYFYLAGNAKSMPADVSEALMSIFQEEGGLCSPDAAAYLARLQQTRRFQTETWA

Specifications

Antigen NADPH dependent diflavin oxidoreductase 1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against a full-length human NDOR1 protein.
Formulation PBS with no preservative; pH 7.4
Gene NDOR1
Gene Accession No. BC015735.1
Gene Alias MGC138148/NR1/bA350O14.9
Gene Symbols NDOR1
Host Species Rabbit
Immunogen NDOR1 (AAH15735.1, 1 a.a. ∼ 597 a.a) full-length human protein.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27158
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.