Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-NDRG1 Clone: 2D7,

Catalog No. AP1160100UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
AP1160100UG 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AP1160100UG Supplier MilliporeSigma™ Supplier No. AP1160100UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifically detects NDRG1 Clone: 2D7 in Human samples, and it is validated for ELISA, Immunocytochemistry, Immunohistochemistry (Paraffin), Immuno Blotting

NDRG1 is a member of the N-myc downregulated gene family which belongs to the alpha/beta hydrolase superfamily. The protein encoded by this gene is a cytoplasmic protein involved in stress responses, hormone responses, cell growth, and differentiation. It is necessary for p53-mediated caspase activation and apoptosis. Mutation in this gene has been reported to be causative for hereditary motor and sensory neuropathy-Lom.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NDRG1
Applications ELISA, Immunoblot, Immunoblot, Immunocytochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 2D7
Conjugate Unconjugated
Formulation In PBS, pH 7.2.
Host Species Mouse
Immunogen Full-length, recombinant human NDRG1 (aa 1-395), expressed as a GST fusion protein
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes the ∽40-45kDa NDRG1 protein in HeLa cells and human endometrium tissue.
Target Species Human
Content And Storage −20°C
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.