Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NDRG2, Mouse, Clone: 1D12, Abnova™

Catalog No. 89002643 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-002-643 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-002-643 Supplier Abnova Corporation Supplier No. H00057447M06
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant NDRG2.

This gene is a member of the N-myc downregulated gene family which belongs to the alpha/beta hydrolase superfamily. The protein encoded by this gene is a cytoplasmic protein that may play a role in neurite outgrowth. This gene may be involved in glioblastoma carcinogenesis. Several alternatively spliced transcript variants of this gene have been described, but the full-length nature of some of these variants has not been determined. [provided by RefSeq

Sequence: MAELQEVQITEEKPLLPGQTPEAAKTHSVETPYGSVTFTVYGTPKPKRPAILTYHDVGLNYKSCFQPLFQFEDMQEIIQNFVRVHVDAPGMEEGAP

Specifications

Antigen NDRG2
Applications ELISA, Western Blot
Classification Monoclonal
Clone 1D12
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant NDRG2.
Formulation PBS with no preservative; pH 7.4
Gene NDRG2
Gene Accession No. NM_016250
Gene Alias DKFZp781G1938/FLJ25522/KIAA1248/SYLD
Gene Symbols NDRG2
Host Species Mouse
Immunogen NDRG2 (NP_057334, 1 a.a. ∼ 96 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57447
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.