Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NDUFA10 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP132146
Description
NDUFA10 Polyclonal specifically detects NDUFA10 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
NDUFA10 | |
Polyclonal | |
Unconjugated | |
0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal | |
NDUFA10 | |
Recombinant protein encompassing a sequence within the center region of human NDUFA10. The exact sequence is proprietary. | |
0.1 mL | |
Primary | |
Tested in human. Not yet tested in any other species. Expected to cross react based on sequence identity: Bovine (82%). | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
0.82mg/mL | |
Western Blot 1:500-1:3000, Immunohistochemistry 10 - 1:500, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000 | |
10 (42kD), CI-42k, CI-42KD, FLJ21923, MGC5103, NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 10, 42kDa, NADH dehydrogenase [ubiquinone] 1 alpha subcomplex subunit 10, mitochondrial | |
Rabbit | |
Affinity Purified | |
RUO | |
4705 | |
Human, Mouse | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction