Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NDUFB10, Rabbit, MaxPab™ Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against a full-length human NDUFB10 protein.
Supplier: Abnova Corporation H00004716D01
Description
Sequence: MPDSWDKDVYPEPPRRTPVQPNPIVYMMKAFDLIVDRPVTLVREFIERQHAKNRYYYYHRQYRRVPDITECKEEDIMCMYEAEMQWKRDYKVDQEIINIMQDRLKACQQREGQNYQQNCIKEVEQFTQVAKAYQDRYQDLGAYSSARKCLAKQRQRMLQERKAAKEAAAATSSpecifications
NDUFB10 | |
Polyclonal | |
Rabbit polyclonal antibody raised against a full-length human NDUFB10 protein. | |
NDUFB10 | |
PDSW | |
Rabbit | |
100 μL | |
Primary | |
Human, Mouse | |
Liquid |
Immunoprecipitation, Western Blot | |
Unconjugated | |
No additive | |
NM_004548.1 | |
NDUFB10 | |
NDUFB10 (NP_004539.1, 1 a.a. ∼ 172 a.a) full-length human protein. | |
RUO | |
4716 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction