Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NADH dehydrogenase (ubiquinone) 1 beta subcomplex, 11, 17.3kDa, Mouse, Clone: 1D6, Abnova™

Catalog No. 89001298 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-001-298 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-001-298 Supplier Abnova Corporation Supplier No. H00054539M12
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a full-length recombinant NDUFB11.

NDUFB11 is a component of mitochondrial complex I. Complex I catalyzes the first step in the electron transport chain, the transfer of 2 electrons from NADH to ubiquinone, coupled to the translocation of 4 protons across the membrane (Carroll et al., 2002 [PubMed 12381726]).[supplied by OMIM

Sequence: MAAGLFGLSARRLLAAAATRGLPAARVRWESSFSRTVVAPSAVAGKRPPEPTTPWQEDPEPEDENLYEKNPDSHGYDKDPVLDVWNMRLVFFFGVSIILVLGSTFVAYLPDYRMKEWSRREAERLVKYREANGLPIMESNCFDPSKIQLPEDE

Specifications

Antigen NADH dehydrogenase (ubiquinone) 1 beta subcomplex, 11, 17.3kDa
Applications ELISA, Western Blot
Classification Monoclonal
Clone 1D6
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a full length recombinant NDUFB11.
Formulation PBS with no preservative; pH 7.4
Gene NDUFB11
Gene Accession No. BC010665
Gene Alias ESSS/FLJ20494/MGC111182/NP17.3/Np15/P17.3
Gene Symbols NDUFB11
Host Species Mouse
Immunogen NDUFB11 (AAH10665, 1 a.a. ∼ 153 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Stem Cell Biology
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 54539
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.