Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NDUFS1, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89003034 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-003-034 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-003-034 Supplier Abnova Corporation Supplier No. H00004719A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length recombinant NDUFS1.

The protein encoded by this gene belongs to the complex I 75kDa subunit family. Mammalian complex I is composed of 45 different subunits. It locates at the mitochondrial inner membrane. This protein has NADH dehydrogenase activity and oxidoreductase activity. It transfers electrons from NADH to the respiratory chain. The immediate electron acceptor for the enzyme is believed to be ubiquinone. This protein is the largest subunit of complex I and it is a component of the iron-sulfur (IP) fragment of the enzyme. It may form part of the active site crevice where NADH is oxidized. Mutations in this gene are associated with complex I deficiency. [provided by RefSeq

Sequence: MLRIPVRKALVGLSKSPKGCVRTTATAASNLIEVFVDGQSVMVEPGTTVLQACEKVGMQIPRFCYHERLSVAGNCRMCLVEIEKAPKVVAACAMPVMKGWNILTNSEKSKKAREGVMEFLLANHPLDCPICDQGGECDLQDQSMMFGNDRSRFLEGKRAVEDKNIGPLVKTIMTRCIQCTRCIRFASEIAGVDDLGTTGRGNDMQVGTYIEKMFMSELSGNIIDICPVGALTSKPYAFTAQPWETRKTESIDVMDAVGSNIVVSTRTGEVMRILPRMHEDINEEWISDKTRFAYDGLKRQRLTEPMVRNEKGLLTYTSWEDALSRVAGMLQSFQGKDVAAIAGGLVDAEALVALKDLLNRVDSDTLCTEEVFPTAGAGTDLRSNYLLNTTIAGVEEADVVLLVGTNPRFEAPLFNARIRKSWLHNDLKVALIGSPVDLTYTYDHLGDSPKILQDIASGSHPFSQVLKEAKKPMVVLGSSALQRNDGAAILAAVSSIAQKIRMTSGVTGDWKVMNILHRIASQVAALDLGYKPGVEAIRKNPPKVLFLLGADGGCITRQDLPKDCFIIYQGHHGDVGAPIADVILPGAAYTEKSATYVNTEGRAQQTKVAVTPPGLAREDWKIIRALSEIAGMTLPYDTLDQVRNRLEEVSPNLVRYDDIEGANYFQQANELSKLVNQQLLADPLVPPQLTIKDFYMTDSISRASQTMAKCVKAVTEGAQAVEEPSIC

Specifications

Antigen NDUFS1
Applications ELISA
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length recombinant NDUFS1.
Formulation 50% glycerol
Gene NDUFS1
Gene Accession No. BC030833
Gene Alias CI-75Kd/MGC26839/PRO1304
Gene Symbols NDUFS1
Host Species Mouse
Immunogen NDUFS1 (AAH30833, 1 a.a. ∼ 727 a.a) full-length recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4719
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.