Learn More
NDUFS2, Rabbit, Monoclonal Antibody, Abnova™
Rabbit monoclonal antibody raised against a human NDUFS2 peptide using ARM Technology.
Supplier: Abnova Corporation H00004720K
Includes: Up to three rabbit IgG clones of 1mg each will be delivered to customer.
Description
Mitochondrial complex I (NADH-ubiquinone reductase; EC 1.6.5.3) is the first multimeric complex of the respiratory chain that catalyzes the NADH oxidation with concomitant ubiquinone reduction and proton ejection out of the mitochondria. Mammalian mitochondrial complex I is an assembly of at least 43 different subunits. Seven of the subunits are encoded by the mitochondrial genome; the remainder are the products of nuclear genes. The iron-sulfur protein (IP) fraction of complex I is made up of 7 subunits, including NDUFS2. See NDUFS1 (MIM 157655).[supplied by OMIM
Library Construction: Nonfusion antibody library from rabbit spleen (ARM Technology)Expression: Overexpression vector and transfection into 293H cell line
Order Info
- Customer may provide cell or tissue lysate for antibody screening.
- Rabbit monoclonal antibody generated by ARM technology is amenable to antibody engineering including F(ab)2, IgG, scFv and different Fc and non-Fc conjugates per customer request.
Specifications
NDUFS2 | |
Monoclonal | |
Unconjugated | |
In 1X PBS, pH 7.4 | |
NDUFS2 | |
Rabbit | |
1 mg x up to 3 Abs | |
Primary | |
Human | |
Liquid |
ELISA, Western Blot | |
1mg | |
NADH dehydrogenase (ubiquinone) Fe-S protein 2, 49kDa (NADH-coenzyme Q reductase) | |
NDUFS2 | |
NDUFS2 | |
A synthetic peptide of human NDUFS2 | |
RUO | |
4720 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.