Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NDUFS2 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA526772
Description
This antibody is predicted to react with bovine and rat based on sequence homology.
NDUFS2 is a core subunit of the mitochondrial membrane respiratory chain NADH dehydrogenase (complex I). Mammalian mitochondrial complex I is composed of at least 43 different subunits, 7 of which are encoded by the mitochondrial genome, and the rest are the products of nuclear genes. The iron-sulfur protein fraction of complex I is made up of 7 subunits, including this gene product. Complex I catalyzes the NADH oxidation with concomitant ubiquinone reduction and proton ejection out of the mitochondria.
Specifications
NDUFS2 | |
Polyclonal | |
NDUFS2 | |
CI-49, CI-49kD, NADH dehydrogenase [ubiquinone] iron-sulfur protein 2, mitochondrial, NADH-ubiquinone oxidoreductase 49 kDa subunit, NADH-ubiquinone oxidoreductase NDUFS2 subunit, complex 1, mitochondrial respiratory chain, 49-KD subunit, complex I 49kDa subunit, comp | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
226646, 4720 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry, Western Blot | |
Unconjugated | |
O75306, Q91WD5 | |
NDUFS2 | |
KLH conjugated synthetic peptide between 286-315 amino acids from the central region of human NDUFS2 | |
400 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction