Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NDUFS7, Mouse, Clone: 3A3, Abnova™

Catalog No. 89002662 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-002-662 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-002-662 Supplier Abnova Corporation Supplier No. H00374291M01
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant NDUFS7.

This gene encodes a protein that is a subunit of one of the complexes that forms the mitochondrial respiratory chain. This protein is one of over 40 subunits found in complex I, the nicotinamide adenine dinucleotide (NADH):ubiquinone oxidoreductase. This complex functions in the transfer of electrons from NADH to the respiratory chain, and ubiquinone is believed to be the immediate electron acceptor for the enzyme. Mutations in this gene cause Leigh syndrome due to mitochondrial complex I deficiency, a severe neurological disorder that results in bilaterally symmetrical necrotic lesions in subcortical brain regions. [provided by RefSeq

Sequence: PRQSDVMIVAGTLTNKMAPALRKVYDQMPEPRYVVSMGSCANGGGYYHYSYSVVRGCDRIVPVDIYIPGCPPTAEALLYGILQLQRKIKRERRLQIWYRR

Specifications

Antigen NDUFS7
Applications ELISA
Classification Monoclonal
Clone 3A3
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant NDUFS7.
Formulation PBS with no preservative; pH 7.4
Gene NDUFS7
Gene Accession No. NM_024407
Gene Alias CI-20KD/FLJ45860/FLJ46880/MGC120002/MY017/PSST
Gene Symbols NDUFS7
Host Species Mouse
Immunogen NDUFS7 (NP_077718, 114 a.a. ∼ 213 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 374291
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.