Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NDUFV3, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89003039 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-003-039 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-003-039 Supplier Abnova Corporation Supplier No. H00004731A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant NDUFV3.

The protein encoded by this gene is one of at least forty-one subunits that make up the NADH-ubiquinone oxidoreductase complex. This complex is part of the mitochondrial respiratory chain and serves to catalyze the rotenone-sensitive oxidation of NADH and the reduction of ubiquinone. The encoded protein is one of three proteins found in the flavoprotein fraction of the complex. The specific function of the encoded protein is unknown. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq

Sequence: MAAPCLLRQGRAGALKTMLQEAQVFRGLASTVSLSAESGKSEKGQPQNSKKQSPPKNVVEPKERGKLLATQTAAELSKNLSSPSSYPPAVNKGRKVASPS

Specifications

Antigen NDUFV3
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant NDUFV3.
Formulation 50% glycerol
Gene NDUFV3
Gene Accession No. NM_021075
Gene Alias CI-9KD
Gene Symbols NDUFV3
Host Species Mouse
Immunogen NDUFV3 (NP_077718, 1 a.a. ∼ 100 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4731
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.