Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NDUFV3, Rabbit, Monoclonal Antibody, Abnova™

Catalog No. 89205832 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mg x up to 3 Abs
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-205-832 1 mg x up to 3 Abs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-205-832 Supplier Abnova Corporation Supplier No. H00004731K

Includes

Up to three rabbit IgG clones of 1mg each will be delivered to customer.

Rabbit monoclonal antibody raised against a human NDUFV3 peptide using ARM Technology.

The protein encoded by this gene is one of at least forty-one subunits that make up the NADH-ubiquinone oxidoreductase complex. This complex is part of the mitochondrial respiratory chain and serves to catalyze the rotenone-sensitive oxidation of NADH and the reduction of ubiquinone. The encoded protein is one of three proteins found in the flavoprotein fraction of the complex. The specific function of the encoded protein is unknown. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq

Library Construction: Nonfusion antibody library from rabbit spleen (ARM Technology)
Expression: Overexpression vector and transfection into 293H cell line

Order Info

  1. Customer may provide cell or tissue lysate for antibody screening.
  2. Rabbit monoclonal antibody generated by ARM technology is amenable to antibody engineering including F(ab)2, IgG, scFv and different Fc and non-Fc conjugates per customer request.

Specifications

Antigen NDUFV3
Applications ELISA, Western Blot
Classification Monoclonal
Concentration 1mg
Conjugate Unconjugated
Description NADH dehydrogenase (ubiquinone) flavoprotein 3, 10kDa
Formulation In 1X PBS, pH 7.4
Gene NDUFV3
Gene Accession No. NDUFV3
Gene Alias CI-9KD
Gene Symbols NDUFV3
Host Species Rabbit
Immunogen A synthetic peptide of human NDUFV3
Quantity 1 mg x up to 3 Abs
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4731
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.