Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Nectin-3/PVRL3 (CD113) Mouse anti-Human, Clone: N3.12.4, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MABT63
Description
Anti-Nectin-3/PVRL3 (CD113) Antibody, clone N3.12.4 detects level of Nectin-3/PVRL3 (CD113) and has been published and validated for use in flow cytometry.
Specifications
Nectin-3/PVRL3 (CD113) | |
Monoclonal | |
Unconjugated | |
NP_056295 | |
Mouse | |
Protein G Purified | |
Primary | |
2-8°C for one year | |
IgG2a κ |
Flow Cytometry | |
N3.12.4 | |
Purified mouse monoclonal IgG2aκ in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide. | |
CDw113, PRR3, PPR3, CD113, PVRR3 | |
Full length recombinant protein corresponding to human Nectin-3. | |
50 μg | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction