Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Nedd4 Rabbit anti-Rat, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma 07049
Description
Anti-Nedd4 Antibody, Cat. No. 07-049, is a rabbit polyclonal antibody that detects Nedd4 and has been tested for use in Immunohistochemistry (Paraffin) and Western Blotting.
Specifications
| Nedd4 | |
| Polyclonal | |
| Purified rabbit polyclonal IgG in buffer containing 0.1M Tris-glycine, pH 7.4, 0.15M NaCl and 0.05% sodium azide before the addition of glycerol to 30%. | |
| Rabbit | |
| Protein A purfied | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Immunohistochemistry, Western Blot | |
| Unconjugated | |
| EC 6.3.2.-; KIAA0093; MGC176705; NEDD-4; NEDD4-1; RPF1 | |
| GST fusion protein corresponding to the WW2 domain (residues 395-462) of rat Nedd4. | |
| 200 μL | |
| NP_006145.2 | |
| -20°C in undiluted aliquots for up to 12 months, Avoid Freeze/Thaw Cycles | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction