Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Neisseria gonorrhoeae, Biotin, Polyclonal, Invitrogen™

Catalog No. PIPA127386
Change view
Click to view available options
Quantity:
100 μL
Catalog No. Quantity
PIPA127386 100 μL
1 options

Catalog No. PIPA127386

Supplier: Thermo Scientific PA127386

Rabbit Polyclonal Antibody

The genus Neisseria contains a number of species which are normal flora and pathogens of humans and animals. Of these species, the species of human origin, and particularly the pathogenic species, N. gonorrhoeae and N. meningitidis, have been studied extensively in an effort to control the infections they cause. Gonorrhea, caused by N. gonorrhoeae, is one of the most frequently reported infectious diseases in the United States and worldwide. Of the Neisseria and related species, only N. gonorrhoeae is considered always to be pathogenic i.e., cause disease. N. gonorrhoeae is not considered to be normal flora under any circumstances.

PA1-27386 detects Neisseria gonorrhoeae from bacteria samples.PA1-27386 has been successfully used in immunofluorescence procedures.The PA1-27386 immunogen is whole Neisseria gonorrhoeae. ATCC 31426
TRUSTED_SUSTAINABILITY

Specifications

Antigen Neisseria gonorrhoeae
Applications ELISA, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Concentration 4mg/mL
Conjugate Biotin
Gene Alias N. gonorrhoeae, Gonococci
Host Species Rabbit
Immunogen Whole Neisseria gonorrhoeae. ATCC 31426
Purification Method IgG fraction
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bacteria
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.