Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NIMA (never in mitosis gene a)-related kinase 6, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89004514 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-004-514 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-004-514 Supplier Abnova Corporation Supplier No. H00010783A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant NEK6.

The Aspergillus nidulans 'never in mitosis A' (NIMA) gene encodes a serine/threonine kinase that controls initiation of mitosis. NIMA-related kinases (NEKs) are a group of protein kinases that are homologous to NIMA. Evidence suggests that NEKs perform functions similar to those of NIMA.[supplied by OMIM

Sequence: MAGQPGHMPHGGSSNNLCHTLGPVHPPDPQRHPNTLSFRCSLADFQIEKKIGRGQFSEVYKATCLLDRKTVALKKVQIFEMMDAKARQDCVKEIGLLKQV

Specifications

Antigen NIMA (never in mitosis gene a)-related kinase 6
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant NEK6.
Formulation 50% glycerol
Gene NEK6
Gene Accession No. BC000101
Gene Alias SID6-1512
Gene Symbols NEK6
Host Species Mouse
Immunogen NEK6 (AAH00101, 1 a.a. ∼ 100 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Research Discipline Kinases
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 10783
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Serum
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.