Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NIMA (never in mitosis gene a)- related kinase 8, Mouse, Purified MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89016639 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-016-639 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-016-639 Supplier Abnova Corporation Supplier No. H00284086B01P
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length human NEK8 protein.

This gene encodes a member of the serine/threionine protein kinase family related to NIMA (never in mitosis, gene A) of Aspergillus nidulans. The encoded protein may play a role in cell cycle progression from G2 to M phase. Mutations in the related mouse gene are associated with a disease phenotype that closely parallels the juvenile autosomal recessive form of polycystic kidney disease in humans. [provided by RefSeq

Sequence: MEKYERIRVVGRGAFGIVHLCLRKADQKLVIIKQIPVEQMTKEERQAAQNECQVLKLLNHPNVIEYYENFLEDKALMIAMEYAPGGTLAEFIQKRCNSLLEEETILHFFVQILLALHHVHTHLILHRDLKTQNILLDKHRMVVKIGDFGISKILSSKSKAYTVVGTPCYISPELCEGKPYNQKSDIWALGCVLYELASLKRAFEAANLPALVLKIMSGTFAPISDRYSPELRQLVLSLLSLEPAQRPPLSHIMAQPLCIRALLNLHTDVGSVRMRRAEKSVAPSNTGSRTTSVRCRGIPRGPVRPAIPPPLSSVYAWGGGLGTPLRLPMLNTEVVQVAAGRTQKAGVTRSGRLILWEAPPLGAGGGSLLPGAVEQPQPQFISRFLEGQSGVTIKHVACGDFFTACLTDRGIIMTFGSGSNGCLGHGSLTDISQPTIVEALLGYEMVQVACGASHVLALSTERELFAWGRGDSGRLGLGTRESHSCPQQVPMPPGQEAQRVVCGIDSSMILTVPGQALACGSNRFNKLGLDHLSLGEEPVPHQQVEEALSFTLLGSAPLDQEPLLSIDLGTAHSAAVTASGDCYTFGSNQHGQLGTNTRRGSRAPCKVQGLEGIKMAMVACGDAFTVAIGAESEVYSWGKGARGRLGRRDEDAGLPRPVQLDETHPYTVTSVSCCHGNTLLAVRSVTDEPVPP

Specifications

Antigen NIMA (never in mitosis gene a)- related kinase 8
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length human NEK8 protein.
Formulation PBS with no preservative; pH 7.4
Gene NEK8
Gene Accession No. NM_178170.2
Gene Alias JCK/MGC138445/NEK12A
Gene Symbols NEK8
Host Species Mouse
Immunogen NEK8 (NP_835464.1, 1 a.a. ∼ 692 a.a) full-length human protein.
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Kinases
Primary or Secondary Primary
Gene ID (Entrez) 284086
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.