Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Neomycin Phosphotransferase II Rabbit anti-Bacteria, Polyclonal, MilliporeSigma™

Catalog No. 06747MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
06-747-MI 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 06-747-MI Supplier MilliporeSigma Supplier No. 06747
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-Neomycin Phosphotransferase II Antibody is an antibody against Neomycin Phosphotransferase II for use in western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Neomycin Phosphotransferase II
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified rabbit polyclonal IgG in buffer containing 0.1M Tris-glycine, pH 7.4, 0.15M NaCl, 0.05% sodium azide before the addition of glycerol to 30%. Liquid at −20°C.
Host Species Rabbit
Immunogen Full length neomycin phosphotransferase (NPT) II
Purification Method Protein A purfied
Quantity 200 μg
Primary or Secondary Primary
Target Species Bacteria
Content And Storage -20°C in undiluted aliquots for up to 12 months, Avoid Freeze/Thaw Cycles
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.