Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Neprilysin/CD10, PE, Polyclonal, R&D Systems™

Catalog No. FAB8160P
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB8160P 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB8160P Supplier R&D Systems Supplier No. FAB8160P
This item is not returnable. View return policy

Host Species: Goat
Isotype: IgG
Species Reactivity: Hu
Immunogen: S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Neprilysin/CD10, Tyr52-Trp750

Applications:

Flow Cytometry

Specifications

Antigen Neprilysin/CD10
Applications Flow Cytometry
Classification Polyclonal
Conjugate R-PE
Dilution Flow Cytometry 10 uL/10^6 cells
Gene Accession No. P08473
Gene Alias Atriopeptidase, CALLA, CALLAmembrane metallo-endopeptidase (neutral endopeptidase, enkephalinase), CD10, CD10 antigen, CD10), CD10membrane metallo-endopeptidase variant 1, Common acute lymphocytic leukemia antigen, DKFZp686O16152, EC 3.4.24, EC 3.4.24.11, Enkephalinase, EPN, Leu-19, membrane metallo-endopeptidase, membrane metallo-endopeptidase variant 2, MGC126681, MGC126707, MME, NEPmembrane metallo-endopeptidase (neutral endopeptidase, enkephalinase, CALLA, neprilysin, Neutral endopeptidase, Neutral Endopeptidase 24.11, NKH1, SFE, Skin fibroblast elastase
Gene Symbols MME
Host Species Goat
Immunogen S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Neprilysin/CD10, Tyr52-Trp750, Accession # P08473
Purification Method Affinity Purified
Quantity 100 Tests
Primary or Secondary Primary
Gene ID (Entrez) 4311
Test Specificity Detects human Neprilysin/CD10 in ELISAs and Western blots. In sandwich ELISAs, approximately 20% cross-reactivity with recombinant mouse Neprilysin is observed, and less than 0.3% cross-reactivity with recombinant human Neprilysin-2 is observed.
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.