Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Neprilysin Rabbit anti-Human, Mouse, Rat, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma AB5458
Description
This Anti-Neprilysin Antibody (Neutral Endopeptidase, Nep) is validated for use in immunohistochemistry, western blotting for the detection of Neprilysin (Neutral Endopeptidase).
Specifications
Neprilysin (Neutral Endopeptidase | |
Polyclonal | |
Rabbit polyclonal serum. Contains no preservative. | |
Rabbit | |
Unpurified | |
Primary | |
Human, Mouse, Rat | |
Serum |
Immunohistochemistry, Western Blot | |
Unconjugated | |
MME; EPN; NEP; MGC126681; CD10; MGC126707; DKFZp686O16152; CALLA; EC 3.4.24.11 | |
Rat Neprilysin. | |
50 μL | |
NM_000902.3 | |
−20°C for up to 12 months |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction