Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nerve Growth Factor-Β Sheep anti-Chicken, Human, Mouse, Rat, Polyclonal, MilliporeSigma™

Catalog No. AB1528SPMI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB1528SPMI 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB1528SPMI Supplier MilliporeSigma Supplier No. AB1528SP
Add to Cart
Add to Cart

Sheep Polyclonal Antibody

Anti-Nerve Growth Factor-β Antibody is an antibody against Nerve Growth Factor-β for use in ELISA, western blot, immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Nerve Growth Factor-β
Applications ELISA, Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Protein G purified immunoglobulin. Lyophilized from PBS, no preservatives. Reconstitute with 500μL of sterile distilled water (1mg/mL).
Gene Symbols NGFB; MGC161426; HSAN5; NGF; Beta-NGF; MGC161428
Host Species Sheep
Immunogen Mouse nerve growth factor-beta purified from salivary glands. Antigen purity greater than 95% by PAGE.
Quantity 500 μg
Primary or Secondary Primary
Gene ID (Entrez) NM_002506.2
Target Species Chicken, Human, Mouse, Rat
Content And Storage −20-−70°C in undiluted aliquotes for 12 months. Avoid repeated freeze/thaw cycles.
Form Purified
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.