Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Nerve Growth Factor-Β Rabbit anti-Chicken, Human, Mouse, Rat, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma AB1526P
Description
This Anti-Nerve Growth Factor-β Antibody, pain is validated for use in ELISA, western blot, immunohistochemistry, FUNC for the detection of Nerve Growth Factor-β.
Specifications
Nerve Growth Factor-β | |
Polyclonal | |
Affinity purified immunoglobulin. Lyophilized from PBS. Contains no preservative. Reconstitute to desired volume with sterile distilled water. Suggested concentration 0.5-1.0mg/mL. | |
Rabbit | |
Affinity Purified | |
Primary | |
Chicken, Human, Mouse, Rat |
ELISA, Immunohistochemistry, Western Blot | |
Unconjugated | |
NGFB; MGC161426; HSAN5; NGF; Beta-NGF; MGC161428 | |
Mouse nerve growth factor-beta purified from salivary glands. | |
20 μg | |
NM_002506.2 | |
Lyophilized: −20-−70°C for up to 12 months. Reconstituted: −20-−70°C in undiluted aliquots for up to 6 months. Avoid repeated freeze/thaw cycles. Avoid repeated freeze/thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction