Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nerve Growth Factor Mouse anti-Bovine, Human, Mouse, Rat, Clone: 27/21, MilliporeSigma™

Catalog No. MAB5260ZMI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB5260ZMI 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB5260ZMI Supplier MilliporeSigma Supplier No. MAB5260Z
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Detect Nerve Growth Factor using this Anti-Nerve Growth Factor Antibody, clone 27/21, azide free validated for use in ELISA, NEUT.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Nerve Growth Factor
Applications ELISA, Neutralization
Classification Monoclonal
Clone 27/21
Conjugate Unconjugated
Formulation Protein A Purified mouse immunoglobulin in 20mM sodium phosphate, 250mM NaCl, pH. 7.6, with no preservatives.
Gene Accession No. NM_001102654.1; NM_002527.4
Gene Symbols NTF3, NT3, NGF2, HDNF, MGC129711, NGF-2, NT-3
Host Species Mouse
Immunogen 2.5 S form of mouse NGF.
Purification Method Protein A purfied
Quantity 25 μg
Primary or Secondary Primary
Target Species Bovine, Human, Mouse, Rat
Content And Storage 2-8°C in undiluted aliquots for up to 6 months. Avoid repeated freeze/thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.