Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nerve Growth Factor Mouse anti-Bovine, Human, Mouse, Rat, Clone: 27/21, MilliporeSigma™

Catalog No. MAB526060UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
60 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB526060UG 60 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB526060UG Supplier MilliporeSigma Supplier No. MAB526060UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This Anti-Nerve Growth Factor Antibody, clone 27/21 is validated for use in ELISA for the detection of Nerve Growth Factor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Nerve Growth Factor
Applications ELISA
Classification Monoclonal
Clone 27/21
Conjugate Unconjugated
Formulation Purified immunoglobulin. Lyophilized from 0.02M Phosphate buffer, 0.25M NaCl with 0.1% sodium azide. Reconstitute with 1 mL of sterile distilled water.
Gene Accession No. NM_001102654.1; NM_002527.4
Gene Symbols NTF3, NT3, NGF2, HDNF, MGC129711, NGF-2, NT-3
Host Species Mouse
Immunogen 2.5 S form of mouse NGF.
Quantity 60 μg
Primary or Secondary Primary
Target Species Bovine, Human, Mouse, Rat
Content And Storage 2-8°C Lyiphilized antibody for up to 12 months. 4°C after resconstitution in unaliquotted but sealed tightly for 6 months; do not freeze thaw.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.