Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nerve Growth Factor Receptor Mouse anti-Rat, Clone: 192-IgG, MilliporeSigma™

Catalog No. MAB365MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB365MI 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB365MI Supplier MilliporeSigma Supplier No. MAB365
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Nerve Growth Factor Receptor Antibody, extracellular, clone 192-IgG is an antibody against Nerve Growth Factor Receptor for use in immunocytochemistry, immunohistochemistry, immunoprecipitation, FUNC and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Nerve Growth Factor Receptor
Applications Functional Assay, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 192-IgG
Conjugate Unconjugated
Formulation Mouse monoclonal IgG1 in 0.02M Phosphate buffer, pH7.6, 0.25M NaCl with 0.1% sodium azide
Gene Accession No. NM_002507.2
Gene Symbols NGFR, p75(NTR), TNFRSF16, Gp80-LNGFR, CD271
Host Species Mouse
Immunogen n-octoglucoside stabilized proteins containing p75 receptor, which were isolated from plasma membranes of PC-12 cells (Chandler et al., 1984).
Purification Method Protein A purfied
Quantity 50 μg
Primary or Secondary Primary
Target Species Rat
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.