Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Nerve Growth Factor Receptor (p75) Mouse anti-Human, Clone: 8211, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MAB5264
Description
Anti-Nerve Growth Factor Receptor (p75) Antibody, clone 8211 is an antibody against Nerve Growth Factor Receptor (p75) for use in immunocytochemistry, immunohistochemistry, immunohistochemistry (paraffin) and western blotting.
Specifications
| Nerve Growth Factor Receptor (p75) | |
| Monoclonal | |
| Unconjugated | |
| NM_002507.2 | |
| Mouse | |
| Protein A purfied | |
| Primary | |
| 2-8°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles. | |
| IgG1 |
| Immunocytochemistry, Immunohistochemistry, Western Blot | |
| 8211 | |
| Protein A Purified mouse immunoglobulin in 20mM sodium phosphate, 250mM NaCl, pH. 7.6, with 0.1% sodium azide as a preservative. | |
| NTF3, NT3, NGF2, HDNF, MGC129711, NGF-2, NT-3 | |
| Membrane fragments of the melanoma cell line WM-9 (Herlyn et al., 1983). | |
| 50 μg | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction