Learn More
MilliporeSigma™ anti-Nerve Growth Factor Receptor, p75 ; Polyclonal; ™ (Chemicon™)
Supplier: MilliporeSigma™ AB1554/DEL
Description
There are two classes of neurotrophin cell-surface receptors. The p75 receptor (also known as the low-affinity neurotrophin receptor, LANR) is common to all members of the neurotrophin family( which includes nerve growth factor (NGF), brain derived neurotrophic factor (BDNF), neurotrophin 3 (NT-3), and neurotrophin 4 (NT-4). The NGF receptor (p75) is belongs to the tumor necrosis factor receptor family and binds all four neurotrophins. The roles of p75 are under investigation but may be involved in regulating the response to the neurotrophins and increasing the affinity of high-affinity Trk receptors (which also bind the four neurotrophins).
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blotting
Specifications
Nerve Growth Factor Receptor (p75) | |
Unconjugated | |
Extracellular fragment generated from sequences from the third exon of mouse p75 (amino acids 43-161). | |
RUO | |
Primary | |
Serum |
Polyclonal | |
Rabbit | |
100 μL | |
Neuroscience | |
Chicken, Human, Mouse, Rat |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.