Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nerve Growth Factor Receptor Rabbit anti-Human, Mouse, Rat, Polyclonal, MilliporeSigma™

Catalog No. AB1554MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB1554MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB1554MI Supplier MilliporeSigma Supplier No. AB1554
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-Nerve Growth Factor Receptor Antibody, p75 is an antibody against Nerve Growth Factor Receptor for use in immunocytochemistry, immunohistochemistry (paraffin),western blotting and immunoprecipitation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Nerve Growth Factor Receptor
Applications Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation
Classification Polyclonal
Conjugate Unconjugated
Formulation Rabbit polyclonal serum containing 0.05% NaN3.
Gene Symbols NGFR; p75(NTR); TNFRSF16; Gp80-LNGFR; CD271
Host Species Rabbit
Immunogen Recombinant mouse NGF Receptor p75 extracellular fragment.
Purification Method Unpurified
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) NP_150086.2
Target Species Human, Mouse, Rat
Content And Storage −20°C for up to 12 months
Form Serum
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.