Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nestin Mouse anti-Human, AlexaFluor™488, Clone: 10C2, MilliporeSigma™

Catalog No. MAB5326A4MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB5326A4MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB5326A4MI Supplier MilliporeSigma Supplier No. MAB5326A4
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Nestin Antibody, clone 10C2, Alexa Fluor 488 conjugate detects level of Nestin and has been published and validated for use in immunocytochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Nestin
Applications Immunocytochemistry
Classification Monoclonal
Clone 10C2
Conjugate Alexa Fluor 488
Formulation Purified mouse monoclonal IgG1 conjugated to Alexa Fluor™ 488 in PBS with 0.1% sodium azide and 15mg/ml BSA.
Gene Accession No. NP_006608
Gene Symbols NES, Nbla00170, FLJ21841, Nestin
Host Species Mouse
Immunogen Fusion protein corresponding to human Nestin.
Purification Method Protein A purified
Quantity 100 μL
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C in undiluted aliquots for up to 6 months. Protect from light.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.