Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nestin Mouse anti-Human, Biotin, Clone: 10C2, MilliporeSigma™

Catalog No. MAB5326BMI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB5326BMI 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB5326BMI Supplier MilliporeSigma Supplier No. MAB5326B

Mouse Monoclonal Antibody

Detect Nestin using this mouse monoclonal antibody, Anti-Nestin Antibody, clone 10C2, Biotin Conjugate validated for use in immunocytochemistry and immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Nestin
Applications Immunocytochemistry, Immunohistochemistry
Classification Monoclonal
Clone 10C2
Conjugate Biotin
Formulation Purified mouse monoclonal IgG1 conjugated to Biotin in PBS with 0.1% sodium azide and 15mg/ml BSA.
Gene Accession No. NP_006608
Gene Symbols NES, Nbla00170, FLJ21841, Nestin.
Host Species Mouse
Immunogen Fusion protein corresponding to human Nestin.
Purification Method Protein A purified
Quantity 50 μL
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C in undiluted aliquots for up to 6 months. Protect from light.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.