Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nestin Mouse, Unlabeled, Clone: 25/NESTIN, BD

Catalog No. BDB611658 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
150 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB611658 50 μg
BDB611659 150 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB611658 Supplier BD Biosciences Supplier No. 611658
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The cytoskeleton consists primarily of core structural proteins that include microfilaments, microtubules, and intermediate filaments (IFs). IFs contain more than 50 distinct proteins that are organized into six different subtypes: Type I/II keratin proteins expressed in epithelia cells, type III vimentin/desmin, type IV neurofilament proteins, type V nuclear lamins, and type VI nestin proteins expressed primarily in embryonic cells. Nestin has a conserved core region (amino acids 7 to 314), which contains an α helical domain that is involved in coiled-coil assembly of IFs. The C-terminal region of nestin is similar to type IV IFs, since it contains highly charged amino acids, many glutamate residues, and an 11 amino acid repeat motif. Nestin is expressed in the cerebrum during embryonic development, in the cerebellum during early postnatal development, and in dermatomal cells and myoblasts during myogenesis. In vitro, nestin forms homodimers and homotetramers, but not IFs, and can co-assemble with type III vimentin and type IV internexin proteins. Thus, nestin is a core IF protein that is essential for proper cytoskeletal formation during neurogenesis and myogenesis. Nestin molecular weight can vary anywhere from 200 to 260kDa depending on where it is being expressed.

Host Species: Mouse
Clone: 25
Isotype: IgG1, κ
Species Reactivity [for Features Main]: Rat
Immunogen: Rat Nestin aa. 402-604

Immunofluorescence, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen Nestin
Applications Western Blot
Classification Monoclonal
Clone 25/NESTIN
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Rat Nestin aa. 402-604 Recombinant Protein
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG1 κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.