Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nestin Mouse anti-Rat, Unlabeled, Clone: RAT 401, BD

Catalog No. BDB556309 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB556309 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB556309 Supplier BD Biosciences Supplier No. 556309
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Multipotential stem cells in the neural tube of the developing embryo give rise to the different neuronal cell types of the brain. Nestin is an intermediate filament protein which is abundantly expressed in neuroepithelial stem cells early in embryogenesis, but is absent from nearly all mature central nervous system (CNS) cells. After its down-regulation, GFAP and neurofilaments are expressed in differentiated astrocytes and neurons, respectively. Initial studies showed that the mouse anti-rat nestin antibody (clone Rat 401) recognized transient radial glial cells and dividing neuroepithelial stem cells in the embryonic rat CNS. Reportedly, Rat 401 has been used to analyze nestin expression in the developing rat nervous system and in immortalized CNS precursor cell lines (e.g such as in E11 rat CNS stem cells, but lost by postnatal day 6 (P6) in spinal cord and by P21 in the cerebellum). Although not expressed in normal adult CNS cells, nestin has been reportedly to be detectable in a variety of CNS tumors, suggesting that these tumors share gene expression patterns with primitive, undifferentiated CNS cells. Rat 401 positive cells have been reported to be found throughout the developing neural tube, but not in the adult CNS. More than 90% of dissociated E11 neural tube cells have been reported to be Rat 401 positive. Rat 401 recognizes rat nestin and has been reported not to cross-react with human nestin. Due to differential tissue expression, this antibody may recognize nestin as a doublet within a range of 198-260kDa.

Host Species: Mouse
Clone: Rat 401
Isotype: IgG1
Species Reactivity: Rat
Immunogen: Rat (E15) spinal cord extracts

Electron Microscopy, Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen Nestin
Applications Western Blot
Classification Monoclonal
Clone RAT 401
Concentration 0.5mg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Immunogen Rat (E15) spinal cord extracts
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Rat
Content And Storage Store undiluted at 4°C
Isotype IgG1
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.