Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nestin Mouse anti-Mouse, Rat, Cy3, Clone: rat-401, MilliporeSigma™

Catalog No. MAB353C3MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB353C3MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB353C3MI Supplier MilliporeSigma Supplier No. MAB353C3
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This Anti-Nestin Antibody, clone rat-401, Cy3 conjugate is validated for use in immunocytochemistry for the detection of Nestin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Nestin
Applications Immunocytochemistry
Classification Monoclonal
Clone rat-401
Conjugate Cyanine3
Formulation Purified mouse monoclonal IgG1 conjugated to Cy3 in PBS with 0.1% sodium azide and 15mg/ml BSA.
Gene Accession No. NP_037119
Gene Symbols NES, Nbla00170, FLJ21841
Host Species Mouse
Immunogen Embryonic rat spinal cord
Purification Method Protein G purified
Quantity 100 μL
Primary or Secondary Primary
Target Species Mouse, Rat
Content And Storage 2-8°C in undiluted aliquots for up to 6 months. Protect from light.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.