Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Netrin receptor UNC5B Rabbit anti-Mouse, Human, Polyclonal, MilliporeSigma™

Catalog No. ABC89MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABC89MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABC89MI Supplier MilliporeSigma Supplier No. ABC89

Rabbit Polyclonal Antibody

Detect Netrin receptor UNC5B using this Anti-Netrin receptor UNC5B Antibody validated for use in western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Netrin receptor UNC5B
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Symbols UNC5B; P53RDL1; UNC5H2
Host Species Rabbit
Immunogen KLH-conjugated branch peptide corresponding to humna Netrin receptor UNC5B.
Purification Method Affinity Purified
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) NP_734465
Target Species Human, Mouse
Content And Storage 2-8°C for 12 months
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.