Learn More
sialidase 1 (lysosomal sialidase), Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant NEU1.
Supplier: Abnova Corporation H00004758A01
Description
The protein encoded by this gene is a lysosomal enzyme that cleaves terminal sialic acid residues from substrates such as glycoproteins and glycolipids. In the lysosome, this enzyme is part of a heterotrimeric complex together with beta-galactosidase and cathepsin A (the latter is also referred to as 'protective protein'). Mutations in this gene can lead to sialidosis, a lysosomal storage disease that can be type 1 (cherry red spot-myoclonus syndrome or normosomatic type), which is late-onset, or type 2 (the dysmorphic type), which occurs at an earlier age with increased severity. [provided by RefSeq]
Sequence: NPAHPEFRVNLTLRWSFSNGTSWRKETVQLWPGPSGYSSLATLEGSMDGEEQAPQLYVLYEKGRNHYTESISVAKISVYGTLSpecifications
sialidase 1 (lysosomal sialidase) | |
Polyclonal | |
Mouse polyclonal antibody raised against a partial recombinant NEU1. | |
NEU1 | |
FLJ93471/NANH/NEU/SIAL1 | |
Mouse | |
50 μL | |
Yes | |
4758 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
ELISA, Western Blot | |
Unconjugated | |
50% glycerol | |
NM_000434 | |
NEU1 | |
NEU1 (NP_000425, 334 a.a. ∼ 415 a.a) partial recombinant protein with GST tag. | |
RUO | |
Primary | |
Human | |
Serum |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.