Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NeuN (rabbit) Rabbit anti-Rat, Mouse, Cy3, Polyclonal, MilliporeSigma™

Catalog No. ABN78C3MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABN78C3MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABN78C3MI Supplier MilliporeSigma Supplier No. ABN78C3
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-NeuN (rabbit) Antibody, Cy3 Conjugate is an antibody against NeuN (rabbit) for use in immunocytochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NeuN
Applications Immunocytochemistry
Classification Polyclonal
Conjugate Cyanine3
Formulation Purified rabbit polyclonal conjugated to Cy3 in PBS with 0.1% sodium azide and 15mg/mL BSA.
Gene Symbols NeuN
Host Species Rabbit
Immunogen GST-tagged recombinant protein corresponding to mouse NeuN.
Purification Method Affinity Purified
Quantity 100 μL
Primary or Secondary Primary
Target Species Mouse, Rat
Content And Storage 2-8°C for 6 months in undiluted aliquotes.
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.