Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NeuN purified Guinea Pig anti-Human, Mouse, Rat, Polyclonal, MilliporeSigma™

Catalog No. ABN90PMI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABN90PMI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABN90PMI Supplier MilliporeSigma Supplier No. ABN90P
Add to Cart
Add to Cart

Guinea Pig Polyclonal Antibody

This Anti-NeuN purified Antibody is validated for use in Western Blotting, immunocytochemistry, immunohistochemistry (paraffin) for the detection of NeuN purified.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NeuN
Applications Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified guinea pig polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Symbols NeuN
Host Species Guinea Pig
Immunogen GST-tagged recombinant protein corresponding to the N-terminus of mouse NeuN.
Purification Method Affinity Purified
Quantity 100 μL
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage 2-8°C for 12 months
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.