Learn More
NeuN (rabbit monoclonal) Rabbit anti-Mouse, Rat, Clone: 27-4, MilliporeSigma™
Rabbit Monoclonal Antibody
Supplier: MilliporeSigma MABN140
Description
Anti-NeuN (rabbit monoclonal) Antibody, clone 27-4 detects level of NeuN (rabbit monoclonal) and has been published and validated for use in immunocytochemistry, Western Blotting and immunohistochemistry.
Specifications
NeuN | |
Monoclonal | |
Unconjugated | |
Rabbit | |
Protein A purified | |
Primary | |
2-8°C for 12 months | |
IgG |
Immunocytochemistry, Immunohistochemistry, Western Blot | |
27-4 | |
Purified Rabbit Monoclonal IgG in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide. | |
Recombinant protein corresponding to mouse NeuN. | |
100 μL | |
Mouse, Rat | |