Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-Neural Cell Adhesion Molecule, Clone: H28.123,

Catalog No. MAB310MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB310MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB310MI Supplier MilliporeSigma™ Supplier No. MAB310

Specifications

Antigen Neural Cell Adhesion Molecule
Applications Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone H28.123
Conjugate Unconjugated
Formulation Purified immunoglobulin. Liquid in PBS containing 1mg/mL BSA. Contains no preservative.
Gene Accession No. NM_000615.5; NM_001076682.2; NM_181351.3
Gene Symbols NCAM1, NCAM-120, CD56, NCAM-140, MSK39, NCAM
Host Species Rat
Immunogen Glycoprotein fraction from neonatal mouse brain
Quantity 100 μg
Primary or Secondary Primary
Target Species Mouse
Content And Storage −20°C in undiluted aliquots for 12 months. Avoid repeated freeze/thaw cycles.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.