Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Neural Cell Adhesion Molecule L1 Rat anti-Mouse, Rat, Clone: 324, MilliporeSigma™
Rat Monoclonal Antibody
Supplier: MilliporeSigma MAB5272
Description
This Anti-Neural Cell Adhesion Molecule L1 Antibody, clone 324 is validated for use in immunocytochemistry, immunohistochemistry, western blotting for the detection of Neural Cell Adhesion Molecule L1.
Specifications
Neural Cell Adhesion Molecule L1 | |
Monoclonal | |
Unconjugated | |
NP_032504.3 | |
Rat | |
Protein A purfied | |
Primary | |
2-8°C for one year | |
IgG |
Immunocytochemistry, Immunohistochemistry, Western Blot | |
324 | |
Rat monoclonal in buffer containing 0.02MPhosphate buffer, 0.25MNaCl, pH 7.6 with 0.1% sodium azide. | |
VCAM1, INCAM-100, MGC99561, CD106, DKFZp779G2333, L1CAM | |
Rats were immunized with a glycoprotein fraction from cerebellum of 8-10 day old C57BL/6J mice. | |
100 μg | |
Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction