Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Neural Cell Adhesion Molecule Rabbit anti-Chicken, Human, Mouse, Rat, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma AB5032
Description
Detect Neural Cell Adhesion Molecule using this Anti-Neural Cell Adhesion Molecule Antibody validated for use in ELISA, immunofluoresence, immunohistochemistry, FUNC and western blotting.
Specifications
Neural Cell Adhesion Molecule | |
Polyclonal | |
Purified rabbit polyclonal in buffer containing PBS containing no preservatives. | |
Rabbit | |
Affinity Purified | |
Primary | |
Chicken, Human, Mouse, Rat |
ELISA, Immunofluorescence, Immunohistochemistry, Western Blot | |
Unconjugated | |
CD56; NCAM; MSK39; NCAM1 | |
Highly purified chicken NCAM | |
50 μg | |
NM_000615.5; NM_001076682.2; NM_181351.3 | |
-20°C in undiluted aliquots for up to 12 months, Avoid Freeze/Thaw Cycles |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction