Learn More
Neurite Outgrowth Inhibitor A Rabbit anti-Human, Mouse, Rat, Polyclonal, MilliporeSigma™
Shop All MilliporeSigma ProductsDescription
Specifications
Specifications
| Antigen | Neurite Outgrowth Inhibitor A |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | Affinity purified immunoglobulin. Liquid in PBS containing 0.1% BSA. |
| Gene Symbols | RTN4; RTN4-B1; Nbla10545; RTN-X; reticulon-4A; Nogo-C; NSP; RTN-x; ASY; foocen; NSP-CL; NOGOC; RTN4-A; Reticulon-4; Nbla00271; KIAA0886; NOGOA; Reticulon-5; Foocen; NI220/250; RTN4-C; RTN4-B2; SP1507; NOGO-A; NOGO; NOGOB; Nogo-B |
| Host Species | Rabbit |
| Immunogen | An 18 amino acid peptide sequence in the middle region of the putative extracellular domain of rat Nogo-A (Chen et al. 2000; GrandPre et al. 2000; Goldberg & Barres 2000; Prinijha et al. 2000; Tessier-Lavigne & Goodman 2000; Nagase et al. 1998). The imm |
| Purification Method | Affinity Purified |
| Quantity | 50 μg |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.